Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 4 | 528 | |||
GenBank mRNA | AK096536.1 | - | 4 | 1683 | ||
AK097462.1 | - | 3 | 386 | |||
BC022559.1 | - | 4 | 527 | |||
BC117490.1 | - | 4 | 490 | |||
BC126474.1 | - | 4 | 490 |