Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 4, 5 | 563 | |||
GenBank mRNA | AK097462.1 | - | 3, 4 | 421 | ||
BC022559.1 | - | 4, 5 | 562 | |||
BC117490.1 | - | 4, 5 | 525 | |||
BC126474.1 | - | 4, 5 | 525 |