Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC093076.1 | - | 2, 3 | 171 | 30414589 | |
BM806224.1 | - | 2, 3 | 160 |