Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178469.3 | NP_848564.2 | 6, 7 | 794 | ||
GenBank mRNA | AK126902.1 | - | 5, 6 | 2347 | ||
AY301268.1 | - | 514 | ||||
BC130465.1 | - | 6, 7 | 749 | 40146496 | ||
BC132714.1 | - | 6, 7 | 749 | 40146736 |