Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026868.1 | 2 | 300 | |||
GenBank mRNA | AK292218.1 | - | 2 | 713 | ||
BC028712.1 | - | 3 | 490 | |||
BC038951.1 | - | 2 | 300 | 5733799 | ||
BC117367.1 | - | 2 | 275 | 40125918 | ||
BC171729.1 | - | 2 | 275 | 9052104 | ||
DB449690.1 | - | 2 | 343 |