Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017930.1 | NP_001017930.1 | 1 | 712 | ||
GenBank mRNA | AK128123.1 | - | 1 | 712 |