Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017931.2 | NP_001017931.2 | 3, 4 | 498 | ||
NM_001171252.1 | NP_001164723.1 | 3, 4 | 512 | |||
NM_001303613.1 | NP_001290542.1 | 3, 4 | 300 | |||
XM_017029282.1 | XP_016884771.1 | 3, 4 | 301 | |||
GenBank mRNA | BC140001.1 | - | 3, 4 | 371 | 8860308 | |
BC140741.1 | - | 3, 4 | 384 | 9021609 | ||
BC144449.1 | - | 3, 4 | 397 | 9052975 | ||
HY049011.1 | - | 3, 4 | 300 |