Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012968.2 | NP_001012986.2 | 1 | 868 | ||
GenBank mRNA | AK126931.1 | - | 1 | 845 | ||
AL833314.1 | - | 1 | 861 | |||
BC043412.1 | - | 1 | 379 | 6156224 | ||
BC112360.1 | - | 1 | 619 | 40032302 |