Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080430.2 | NP_536355.1 | 3 | 246 | ||
GenBank mRNA | AY043487.1 | - | 3 | 275 | ||
BC013421.1 | - | 3 | 192 | 3890282 | ||
BC030236.1 | - | 3 | 203 | 4184342 | ||
BC068004.1 | - | 3 | 236 | 6058758 |