Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other XRRA1 SIRNA Assays ›
Gene Symbol:
XRRA1
Gene Name:
X-ray radiation resistance associated 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 74840910 - 74949187 on Build GRCh38
UniGene ID:
Hs.370145
Species:
Human
siRNA ID
HSS152056
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: XRRA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270380.1
NP_001257309.1
12
1692
NM_001270381.1
NP_001257310.1
12
1685
NM_182969.2
NP_892014.1
15
1953
XM_005273765.2
XP_005273822.1
15
1977
XM_005273770.2
XP_005273827.1
13
1757
XM_011544755.1
XP_011543057.1
15
1932
XM_011544756.1
XP_011543058.1
14
1851
XM_011544757.1
XP_011543059.1
14
1806
XM_011544758.1
XP_011543060.1
13
1806
XM_011544759.1
XP_011543061.1
13
1782
XM_011544760.2
XP_011543062.1
15
1977
XM_011544761.1
XP_011543063.1
12
1734
XM_011544762.1
XP_011543064.1
12
1716
XM_011544763.1
XP_011543065.1
15
1977
XM_011544764.1
XP_011543066.1
12
1641
XM_011544765.1
XP_011543067.1
13
1691
XM_011544766.2
XP_011543068.1
15
1996
XM_011544767.1
XP_011543069.1
14
1898
XM_011544768.1
XP_011543070.1
13
1745
XM_017017221.1
XP_016872710.1
13
1767
XM_017017222.1
XP_016872711.1
14
1851
XM_017017223.1
XP_016872712.1
13
1806
XM_017017224.1
XP_016872713.1
12
1716
XM_017017225.1
XP_016872714.1
12
1641
XM_017017226.1
XP_016872715.1
14
1872
XM_017017227.1
XP_016872716.1
14
1848
XM_017017228.1
XP_016872717.1
13
1757
XM_017017229.1
XP_016872718.1
13
1701
XM_017017230.1
XP_016872719.1
12
1525
XM_017017231.1
XP_016872720.1
11
1611
XM_017017232.1
XP_016872721.1
10
1421
XM_017017233.1
XP_016872722.1
12
1705
XM_017017234.1
XP_016872723.1
11
1590
GenBank mRNA
AF439934.1
-
4
578
AK056364.1
-
10
1276
AK074152.1
-
10
1283
AK091449.1
-
2
1399
BC037294.2
-
12
1618
BC040541.1
-
12
1618
BC064584.1
-
12
1621
5742492
CR749605.1
-
11
1657
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to X-ray
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-zrdb5:80/100.66.79.80:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline