Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152432.2 | NP_689645.2 | 7 | 692 | ||
XM_011542615.2 | XP_011540917.1 | 7 | 3180 | |||
XM_011542616.2 | XP_011540918.1 | 7 | 949 | |||
XM_011542617.2 | XP_011540919.1 | 8 | 835 | |||
XM_017017237.1 | XP_016872726.1 | 7 | 1138 | |||
XM_017017238.1 | XP_016872727.1 | 8 | 3321 | |||
GenBank mRNA | DA800121.1 | - | 3 | 257 |