Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2CD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207322.2 | NP_997205.2 | 2 | 169 | ||
GenBank mRNA | AF504646.1 | - | 2 | 143 | ||
BM742013.1 | - | 2 | 169 |