Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145646.1 | NP_001139118.1 | 5 | 3649 | ||
NM_031301.3 | NP_112591.2 | 6 | 3772 | |||
GenBank mRNA | AK057677.1 | - | 1 | 2170 |