Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138573.3 | NP_612640.1 | 4 | 313 | ||
XM_017021937.1 | XP_016877426.1 | 8 | 824 | |||
XM_017021938.1 | XP_016877427.1 | 7 | 3031 | |||
XM_017021939.1 | XP_016877428.1 | 7 | 593 | |||
XM_017021940.1 | XP_016877429.1 | 6 | 3369 | |||
XM_017021941.1 | XP_016877430.1 | 4 | 332 | |||
XM_017021942.1 | XP_016877431.1 | 4 | 350 | |||
XM_017021943.1 | XP_016877432.1 | 4 | 423 | |||
XM_017021944.1 | XP_016877433.1 | 7 | 3416 | |||
XM_017021945.1 | XP_016877434.1 | 7 | 3408 | |||
XM_017021946.1 | XP_016877435.1 | 7 | 3408 | |||
XM_017021947.1 | XP_016877436.1 | 7 | 3408 | |||
XM_017021948.1 | XP_016877437.1 | 5 | 3141 | |||
GenBank mRNA | AK311204.1 | - | 9 | 645 | ||
AK311507.1 | - | 5 | 415 | |||
AM392364.1 | - | 2 | 131 | |||
BC017568.1 | - | 4 | 317 | 4071605 | ||
BX538100.1 | - | 5 | 343 | |||
CR749519.1 | - | 6 | 487 |