Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 31, 32 | 3515 | ||
XM_011545742.2 | XP_011544044.1 | 35, 36 | 3797 | |||
XM_011545744.2 | XP_011544046.1 | 20, 21 | 2397 | |||
XM_011545745.1 | XP_011544047.1 | 18, 19 | 2165 | |||
XM_011545746.2 | XP_011544048.1 | 18, 19 | 2268 | |||
XM_017022947.1 | XP_016878436.1 | 36, 37 | 3870 | |||
XM_017022948.1 | XP_016878437.1 | 19, 20 | 2969 | |||
XM_017022949.1 | XP_016878438.1 | 17, 18 | 3240 | |||
GenBank mRNA | AK093467.1 | - | 7, 8 | 1078 | ||
AK128606.1 | - | 20, 21 | 2383 | |||
AY360464.1 | - | 6, 7 | 721 | |||
BC038400.1 | - | 8, 9 | 960 | |||
BC109294.1 | - | 20, 21 | 2290 | |||
BC109295.1 | - | 19, 20 | 2243 |