Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305203.1 | NP_001292132.1 | 5 | 1637 | ||
NM_001305206.1 | NP_001292135.1 | 5 | 1692 | |||
NM_001305207.1 | NP_001292136.1 | 5 | 1689 | |||
NM_001305208.1 | NP_001292137.1 | 5 | 1682 | |||
NM_133458.3 | NP_597715.2 | 5 | 1627 | |||
NR_130976.1 | 5 | 1735 | ||||
NR_130977.1 | 5 | 1685 | ||||
NR_130978.1 | 5 | 1743 | ||||
XM_005255804.2 | XP_005255861.1 | 5 | 1567 | |||
XM_017022952.1 | XP_016878441.1 | 4 | 1674 | |||
GenBank mRNA | AB075834.1 | - | 1 | 831 | ||
AK056596.1 | - | 1 | 420 | |||
AK074332.1 | - | 5 | 1651 | |||
AK122847.1 | - | 1 | 2062 | |||
AK304152.1 | - | 5 | 1529 | |||
AK316251.1 | - | 5 | 1554 | |||
BC033165.1 | - | 1 | 324 | |||
BC040486.1 | - | 5 | 1637 | |||
BC140886.1 | - | 5 | 1627 | 9021754 | ||
BC140887.1 | - | 5 | 1627 | 9021755 |