Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103175.1 | NP_001096645.1 | 5 | 933 | ||
XM_005255135.4 | XP_005255192.1 | 6 | 994 | |||
XM_011522391.1 | XP_011520693.1 | 5 | 1182 | |||
GenBank mRNA | AL833717.1 | - | 3 | 2139 | ||
AL833749.1 | - | 4 | 2458 | |||
BC128602.1 | - | 5 | 933 | 40132962 |