Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027274.1 | 1, 2 | 346 | |||
GenBank mRNA | AK098626.1 | - | 1, 2 | 346 |