Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027274.1 | 2 | 430 | |||
NR_027275.1 | 3 | 361 | ||||
GenBank mRNA | AK056063.1 | - | 2 | 1352 | ||
DB448439.2 | - | 3 | 361 |