Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010855.3 | NP_001010855.1 | 11 | 1497 | ||
NM_001290211.1 | NP_001277140.1 | 11 | 1865 | |||
NR_110865.1 | 10 | 1592 | ||||
XM_011523677.2 | XP_011521979.1 | 12 | 1510 | |||
XM_011523678.2 | XP_011521980.1 | 12 | 1494 | |||
XM_011523680.2 | XP_011521982.1 | 12 | 1510 | |||
XM_011523681.2 | XP_011521983.1 | 11 | 1758 | |||
XM_011523683.2 | XP_011521985.1 | 9 | 1195 | |||
XM_011523686.2 | XP_011521988.1 | 3 | 644 | |||
XM_017024228.1 | XP_016879717.1 | 12 | 1509 | |||
XM_017024229.1 | XP_016879718.1 | 12 | 1509 | |||
XM_017024230.1 | XP_016879719.1 | 12 | 1509 | |||
XM_017024231.1 | XP_016879720.1 | 8 | 1686 | |||
GenBank mRNA | AK091819.1 | - | 10 | 1594 | ||
AY753192.1 | - | 9 | 1257 | |||
BC110301.1 | - | 11 | 1848 | |||
BC125169.1 | - | 6 | 901 |