Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145272.3 | NP_660315.2 | 3 | 453 | ||
GenBank mRNA | AY557347.1 | - | 2 | 408 | ||
DB452451.1 | - | 3 | 364 |