Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026899.1 | 2 | 197 | |||
GenBank mRNA | AK055342.1 | - | 1 | 68 | ||
BC120976.1 | - | 1 | 149 | |||
BC120977.2 | - | 1 | 149 | |||
DB093141.1 | - | 2 | 197 |