Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026899.1 | 3 | 832 | |||
NR_027487.2 | 2 | 620 | ||||
GenBank mRNA | AK055342.1 | - | 2 | 703 | ||
BC032382.1 | - | 2 | 620 | |||
BC120976.1 | - | 2 | 784 | |||
BC120977.2 | - | 2 | 784 |