Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264573.1 | NP_001251503.1 | 14 | 2473 | ||
NM_001265577.1 | NP_001252506.1 | 14 | 2437 | |||
XM_011524385.2 | XP_011522687.1 | 14 | 2774 | |||
XM_011524386.2 | XP_011522688.1 | 14 | 2508 | |||
XM_011524387.2 | XP_011522689.1 | 14 | 2748 | |||
XM_011524388.2 | XP_011522690.1 | 14 | 2738 | |||
XM_011524389.2 | XP_011522691.1 | 14 | 2774 | |||
XM_011524390.2 | XP_011522692.1 | 13 | 2670 | |||
GenBank mRNA | BC044933.1 | - | 5 | 1106 | ||
BC067365.1 | - | 2 | 636 | |||
BC136590.1 | - | 13 | 2363 | 9020580 | ||
BC144271.1 | - | 13 | 2399 | 9052791 | ||
GU808359.1 | - | 13 | 2402 |