Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRHR1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026905.2 | 2 | 1957 | |||
GenBank mRNA | AK057897.1 | - | 2 | 717 | ||
AK090656.1 | - | 1 | 1436 | |||
BC069230.1 | - | 2 | 1678 | |||
BU684887.1 | - | 1 | 517 |