Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178493.5 | NP_848588.3 | 9, 10 | 1404 | ||
GenBank mRNA | AB062438.1 | - | 9, 10 | 830 | ||
BC036872.1 | - | 9, 10 | 953 | 5225889 | ||
BC060882.1 | - | 9, 10 | 1412 | 30336041 |