Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182919.3 | NP_891549.1 | 2 | 1693 | ||
GenBank mRNA | AB086380.1 | - | 1 | 1487 | ||
AB093555.1 | - | 1 | 1425 | |||
AB097023.1 | - | 1 | 1425 | |||
AB446484.1 | - | 1 | 1425 | |||
AF070530.1 | - | 1 | 539 | |||
AF492646.1 | - | 1 | 1013 | |||
BC009860.2 | - | 1 | 359 | |||
BC136556.1 | - | 1 | 1479 | 9020546 | ||
BC136557.1 | - | 1 | 1479 | 9020547 |