Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 12 | 1717 | ||
GenBank mRNA | AK054643.1 | - | 6 | 1119 | ||
BC024295.2 | - | 8 | 1292 | 3354502 | ||
BC033213.1 | - | 6 | 1082 | 5014368 | ||
JX093066.1 | - | 11 | 1592 | |||
JX093067.1 | - | 11 | 1640 | |||
JX093068.1 | - | 11 | 1640 | |||
JX093070.1 | - | 11 | 1592 | |||
JX093071.1 | - | 11 | 1589 | |||
JX093072.1 | - | 8 | 1262 | |||
JX093073.1 | - | 11 | 1640 | |||
JX093074.1 | - | 11 | 1559 | |||
JX093075.1 | - | 11, 12 | 1640 | |||
JX093076.1 | - | 10 | 1451 | |||
JX093077.1 | - | 10 | 1397 | |||
JX093078.1 | - | 10 | 1831 | |||
JX093079.1 | - | 9 | 1313 | |||
JX093080.1 | - | 10 | 1786 | |||
JX093081.1 | - | 9 | 1310 | |||
JX093082.1 | - | 10 | 1783 | |||
JX093083.1 | - | 10 | 1783 | |||
JX093084.1 | - | 8 | 1507 | |||
JX093086.1 | - | 11 | 1566 | |||
JX093087.1 | - | 11 | 1506 | |||
JX093089.1 | - | 11 | 1561 | |||
JX093090.1 | - | 11 | 1555 | |||
JX093093.1 | - | 10 | 1474 | |||
JX093094.1 | - | 10 | 1429 | |||
JX093095.1 | - | 10 | 1426 | |||
JX093096.1 | - | 9 | 1242 | |||
JX093097.1 | - | 8 | 1126 | |||
JX093098.1 | - | 5 | 1151 | |||
JX093099.1 | - | 11 | 1919 | |||
JX093100.1 | - | 12 | 1722 | |||
JX093101.1 | - | 11 | 1514 | |||
JX093102.1 | - | 5 | 1027 | |||
JX093103.1 | - | 11 | 1626 | |||
JX093104.1 | - | 8 | 1728 | |||
JX093105.1 | - | 10 | 1515 | |||
JX093106.1 | - | 9 | 1301 | |||
JX093107.1 | - | 8 | 1185 | |||
JX093108.1 | - | 10 | 1464 | |||
JX093109.1 | - | 7 | 1077 | |||
JX093110.1 | - | 8 | 1244 |