Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 12, 13 | 1760 | ||
GenBank mRNA | AK054643.1 | - | 6, 7 | 1162 | ||
BC024295.2 | - | 8, 9 | 1335 | 3354502 | ||
BC033213.1 | - | 6, 7 | 1125 | 5014368 | ||
JX093066.1 | - | 11, 12 | 1635 | |||
JX093067.1 | - | 11, 12 | 1683 | |||
JX093068.1 | - | 11, 12 | 1683 | |||
JX093070.1 | - | 11, 12 | 1635 | |||
JX093071.1 | - | 11, 12 | 1632 | |||
JX093072.1 | - | 8, 9 | 1305 | |||
JX093073.1 | - | 11, 12 | 1683 | |||
JX093074.1 | - | 11, 12 | 1602 | |||
JX093076.1 | - | 10, 11 | 1494 | |||
JX093077.1 | - | 10, 11 | 1440 | |||
JX093078.1 | - | 10, 11 | 1874 | |||
JX093079.1 | - | 9, 10 | 1356 | |||
JX093080.1 | - | 10, 11 | 1829 | |||
JX093081.1 | - | 9, 10 | 1353 | |||
JX093082.1 | - | 10, 11 | 1826 | |||
JX093083.1 | - | 10, 11 | 1826 | |||
JX093084.1 | - | 8, 9 | 1550 | |||
JX093086.1 | - | 11, 12 | 1609 | |||
JX093087.1 | - | 11, 12 | 1549 | |||
JX093089.1 | - | 11, 12 | 1604 | |||
JX093090.1 | - | 11, 12 | 1598 | |||
JX093093.1 | - | 10, 11 | 1517 | |||
JX093094.1 | - | 10, 11 | 1472 | |||
JX093095.1 | - | 10, 11 | 1469 | |||
JX093096.1 | - | 9, 10 | 1285 | |||
JX093097.1 | - | 8, 9 | 1169 | |||
JX093098.1 | - | 5, 6 | 1194 | |||
JX093099.1 | - | 11, 12 | 1962 | |||
JX093100.1 | - | 12, 13 | 1765 | |||
JX093101.1 | - | 11, 12 | 1557 | |||
JX093102.1 | - | 5, 6 | 1070 | |||
JX093103.1 | - | 11, 12 | 1669 | |||
JX093104.1 | - | 8, 9 | 1771 | |||
JX093105.1 | - | 10, 11 | 1558 | |||
JX093106.1 | - | 9, 10 | 1344 | |||
JX093107.1 | - | 8, 9 | 1228 | |||
JX093108.1 | - | 10, 11 | 1507 | |||
JX093109.1 | - | 7, 8 | 1120 | |||
JX093110.1 | - | 8, 9 | 1287 |