Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 14 | 2079 | ||
GenBank mRNA | AK054643.1 | - | 7 | 1580 | ||
BC024295.2 | - | 10 | 1654 | 3354502 | ||
BC033213.1 | - | 8 | 1444 | 5014368 | ||
BM055062.1 | - | 1 | 472 | |||
JX093066.1 | - | 12 | 2053 | |||
JX093067.1 | - | 13 | 2017 | |||
JX093068.1 | - | 13 | 2002 | |||
JX093069.1 | - | 12 | 1826 | |||
JX093070.1 | - | 13 | 1954 | |||
JX093071.1 | - | 13 | 1951 | |||
JX093072.1 | - | 10 | 1624 | |||
JX093073.1 | - | 13 | 1939 | |||
JX093074.1 | - | 13 | 1921 | |||
JX093075.1 | - | 12 | 1813 | |||
JX093076.1 | - | 12 | 1813 | |||
JX093077.1 | - | 12 | 1759 | |||
JX093078.1 | - | 12 | 2193 | |||
JX093079.1 | - | 11 | 1675 | |||
JX093080.1 | - | 11 | 2247 | |||
JX093081.1 | - | 11 | 1672 | |||
JX093082.1 | - | 12 | 2145 | |||
JX093083.1 | - | 11 | 2244 | |||
JX093084.1 | - | 10 | 1869 | |||
JX093085.1 | - | 8 | 971 | |||
JX093086.1 | - | 13 | 1928 | |||
JX093087.1 | - | 13 | 1868 | |||
JX093088.1 | - | 8 | 968 | |||
JX093089.1 | - | 13 | 1923 | |||
JX093090.1 | - | 13 | 1917 | |||
JX093091.1 | - | 5 | 894 | |||
JX093092.1 | - | 10 | 1018 | |||
JX093093.1 | - | 12 | 1836 | |||
JX093094.1 | - | 12 | 1791 | |||
JX093095.1 | - | 12 | 1788 | |||
JX093096.1 | - | 11 | 1604 | |||
JX093097.1 | - | 10 | 1488 | |||
JX093098.1 | - | 7 | 1513 | |||
JX093099.1 | - | 13 | 2281 | |||
JX093100.1 | - | 14 | 2084 | |||
JX093101.1 | - | 13 | 1876 | |||
JX093102.1 | - | 7 | 1389 | |||
JX093103.1 | - | 13 | 1988 | |||
JX093104.1 | - | 10 | 2090 | |||
JX093105.1 | - | 12 | 1877 | |||
JX093106.1 | - | 11 | 1663 | |||
JX093107.1 | - | 10 | 1547 | |||
JX093108.1 | - | 12 | 1826 | |||
JX093109.1 | - | 9 | 1439 | |||
JX093110.1 | - | 10 | 1606 |