Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173509.2 | NP_775780.1 | 4 | 416 | ||
XM_005244913.2 | XP_005244970.1 | 5 | 853 | |||
XM_006711174.2 | XP_006711237.1 | 5 | 564 | |||
XM_006711175.2 | XP_006711238.1 | 6 | 676 | |||
XM_006711176.2 | XP_006711239.1 | 4 | 542 | |||
XM_006711177.1 | XP_006711240.1 | 5 | 396 | |||
XM_011509217.1 | XP_011507519.1 | 7 | 698 | |||
XM_011509221.1 | XP_011507523.1 | 6 | 504 | |||
XM_011509226.1 | XP_011507528.1 | 4 | 338 | |||
XM_011509228.1 | XP_011507530.1 | 4 | 472 | |||
XM_017000386.1 | XP_016855875.1 | 7 | 847 | |||
XM_017000387.1 | XP_016855876.1 | 8 | 969 | |||
XM_017000388.1 | XP_016855877.1 | 6 | 710 | |||
XM_017000389.1 | XP_016855878.1 | 8 | 981 | |||
XM_017000390.1 | XP_016855879.1 | 6 | 701 | |||
XM_017000391.1 | XP_016855880.1 | 6 | 825 | |||
XM_017000392.1 | XP_016855881.1 | 5 | 350 | |||
XM_017000393.1 | XP_016855882.1 | 4 | 209 | |||
GenBank mRNA | AB299025.1 | - | 1 | 20 | ||
AK292432.1 | - | 5 | 480 | |||
AK310711.1 | - | 4 | 472 | |||
AK313887.1 | - | 4 | 223 | |||
AY902195.1 | - | 4 | 416 | |||
BC009382.2 | - | 4 | 416 | 4124490 |