Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136537.1 | NP_001130009.1 | 6 | 901 | ||
XM_006710386.3 | XP_006710449.1 | 6 | 1939 | |||
XM_011540803.2 | XP_011539105.1 | 5 | 2201 | |||
XM_017000447.1 | XP_016855936.1 | 6 | 2235 | |||
XM_017000448.1 | XP_016855937.1 | 7 | 1972 | |||
XM_017000449.1 | XP_016855938.1 | 6 | 2234 | |||
GenBank mRNA | AK056150.1 | - | 2 | 532 | ||
AK304611.1 | - | 7 | 885 |