Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120595.1 | 2 | 958 | |||
NR_120596.1 | 2 | 539 | ||||
NR_120597.1 | 2 | 196 | ||||
GenBank mRNA | AI914073.1 | - | 1 | 285 | ||
AV709904.1 | - | 2 | 530 | |||
BC038245.1 | - | 2 | 924 | |||
CB050274.1 | - | 2 | 205 |