Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277074.1 | NP_001264003.1 | 1 | 96 | ||
NM_144716.5 | NP_653317.2 | 2 | 251 | |||
XM_011533391.2 | XP_011531693.1 | 2 | 257 | |||
XM_011533392.2 | XP_011531694.1 | 2 | 209 | |||
GenBank mRNA | AK096749.1 | - | 1 | 74 | ||
AK295532.1 | - | 1 | 74 | |||
AK303160.1 | - | 1 | 88 | |||
BC020830.1 | - | 1 | 77 | 4771328 | ||
BQ709657.1 | - | 2 | 215 | |||
CN430023.1 | - | 2 | 206 | |||
CV574708.1 | - | 3 | 327 | |||
DC379144.1 | - | 1 | 99 |