Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099777.3 | NP_001093247.1 | 2 | 262 | ||
NM_001130001.2 | NP_001123473.1 | 2 | 262 | |||
NM_001130002.2 | NP_001123474.1 | 2 | 262 | |||
NM_001243729.1 | NP_001230658.1 | 2 | 262 | |||
NR_024016.2 | 2 | 262 | ||||
XM_017005754.1 | XP_016861243.1 | 2 | 216 | |||
GenBank mRNA | AA131625.1 | - | 2 | 238 | ||
BC010062.2 | - | 2 | 162 | |||
BC029129.1 | - | 2 | 209 | |||
BC126187.1 | - | 2 | 229 | 8991903 | ||
BC126189.1 | - | 2 | 229 | 8991905 | ||
BC144066.1 | - | 2 | 229 | |||
BE905784.1 | - | 2 | 262 | |||
BG217991.1 | - | 1 | 96 | |||
BX113701.1 | - | 2 | 233 |