Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083308.1 | NP_001076777.1 | 1 | 139 | ||
GenBank mRNA | AY858112.1 | - | 1 | 139 |