Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT16P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002949.2 | 2 | 311 | |||
NR_027766.1 | 1 | 385 | ||||
GenBank mRNA | AK000532.1 | - | 2 | 262 | ||
AK297126.1 | - | 2 | 311 | |||
BC015593.2 | - | 1 | 347 | |||
DA541046.1 | - | 1 | 385 |