Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009555.3 | NP_001009555.3 | 7 | 1561 | ||
NM_001128923.1 | NP_001122395.1 | 7 | 1549 | |||
NM_001128924.1 | NP_001122396.1 | 1 | 660 | |||
NM_001243349.1 | NP_001230278.1 | 7 | 1470 | |||
XM_005262767.2 | XP_005262824.1 | 8 | 1422 | |||
XM_011531646.1 | XP_011529948.1 | 6 | 1208 | |||
XM_011531647.1 | XP_011529949.1 | 6 | 1208 | |||
XM_011531648.1 | XP_011529950.1 | 8 | 1535 | |||
XM_011531649.1 | XP_011529951.1 | 6 | 1208 | |||
XM_017007778.1 | XP_016863267.1 | 8 | 1526 | |||
XM_017007779.1 | XP_016863268.1 | 6 | 1199 | |||
XM_017007780.1 | XP_016863269.1 | 8 | 1526 | |||
XM_017007781.1 | XP_016863270.1 | 7 | 1452 | |||
XM_017007782.1 | XP_016863271.1 | 7 | 1452 | |||
XM_017007783.1 | XP_016863272.1 | 8 | 1422 | |||
GenBank mRNA | AK294476.1 | - | 7 | 1470 | ||
BC108890.1 | - | 1 | 660 | 40001764 | ||
BC108891.1 | - | 1 | 660 | 40001765 | ||
BC108892.1 | - | 1 | 660 | 40001766 | ||
BC108893.1 | - | 1 | 660 | 40001767 | ||
BX647422.1 | - | 7 | 1473 |