Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206981.1 | NP_001193910.1 | 3 | 361 | ||
NM_178497.3 | NP_848592.2 | 2 | 317 | |||
XM_011531668.2 | XP_011529970.1 | 4 | 1316 | |||
GenBank mRNA | AK074237.1 | - | 2 | 317 | ||
AK172776.1 | - | 2 | 317 | |||
AK300227.1 | - | 3 | 344 | |||
BC117342.1 | - | 2 | 317 | 40125893 |