Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBLAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203406.1 | NP_981951.1 | 1 | 508 | ||
GenBank mRNA | AK096942.1 | - | 1 | 118 | ||
AK289778.1 | - | 1 | 60 | |||
BC038230.1 | - | 1 | 508 | 5729339 | ||
BI918374.1 | - | 1 | 152 |
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543100.2 | XP_011541402.1 | 1 | 329 |