Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038216.1 | 2, 3 | 494 | |||
NR_038217.1 | 2, 3 | 285 | ||||
NR_038218.1 | 2, 3 | 251 | ||||
NR_038219.1 | 4, 5 | 464 | ||||
GenBank mRNA | AK055166.1 | - | 2, 3 | 288 | ||
BC041589.1 | - | 2, 3 | 223 | |||
DA244667.1 | - | 2, 3 | 240 | |||
DA265202.1 | - | 2, 3 | 462 | |||
DA291077.1 | - | 4, 5 | 452 | |||
DA419376.1 | - | 2, 3 | 288 | |||
DA420003.1 | - | 2, 3 | 496 |