Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038216.1 | 3 | 529 | |||
NR_038217.1 | 3 | 320 | ||||
NR_038218.1 | 3 | 286 | ||||
NR_038219.1 | 5 | 499 | ||||
GenBank mRNA | AK055166.1 | - | 3 | 323 | ||
BC041589.1 | - | 3 | 258 | |||
DA244667.1 | - | 3 | 275 | |||
DA291077.1 | - | 5 | 487 | |||
DA419376.1 | - | 3 | 323 | |||
DA420003.1 | - | 3 | 531 |