Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278589.1 | NP_001265518.1 | 9 | 1198 | ||
NM_003932.4 | NP_003923.2 | 9 | 1228 | |||
GenBank mRNA | AK130770.1 | - | 9 | 861 | ||
AK303530.1 | - | 9 | 829 | |||
AK312399.1 | - | 9 | 843 | |||
AK316155.1 | - | 9 | 747 | |||
AY513286.1 | - | 9 | 826 | |||
AY826824.1 | - | 9 | 837 | |||
AY826825.1 | - | 9 | 837 | |||
BC052982.1 | - | 9 | 850 | 6046993 | ||
BC071629.1 | - | 9 | 828 | 4536907 | ||
BC121107.1 | - | 9 | 836 | |||
BC121108.1 | - | 9 | 836 | |||
BC139724.1 | - | 9 | 785 | 40031942 | ||
CR456586.1 | - | 9 | 868 | |||
U17714.1 | - | 9 | 834 | |||
U28918.1 | - | 9 | 853 |
Gene Symbol: ST13P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002183.1 | 1 | 819 | |||
GenBank mRNA | AF539468.1 | - | 1 | 819 |