Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176813.4 | NP_789783.1 | 2 | 94 | ||
XM_005249627.2 | XP_005249684.1 | 2 | 73 | |||
XM_011515152.2 | XP_011513454.1 | 2 | 86 | |||
GenBank mRNA | AY069977.1 | - | 1 | 59 | ||
AY359065.1 | - | 2 | 78 | |||
BC058284.1 | - | 2 | 99 | 4694757 | ||
BG540617.1 | - | 96 |