Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145283.2 | NP_660326.2 | 1, 2 | 640 | ||
GenBank mRNA | BC022521.1 | - | 1, 2 | 636 | 4793993 |