Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AWAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013579.2 | NP_001013597.1 | 2 | 170 | ||
XM_017029295.1 | XP_016884784.1 | 2 | 1336 | |||
GenBank mRNA | AY947638.1 | - | 2 | 129 | ||
W68831.1 | - | 2 | 170 |