Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2BFWT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002916.4 | NP_001002916.3 | 1, 2 | 506 | ||
GenBank mRNA | AY283370.1 | - | 1, 2 | 479 | ||
BC038109.2 | - | 1, 2 | 503 | 5266336 | ||
BC118604.1 | - | 1, 2 | 499 | 40109007 | ||
BC121816.1 | - | 1, 2 | 499 | 40109006 |