Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2BFWT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002916.4 | NP_001002916.3 | 2, 3 | 570 | ||
GenBank mRNA | BC038109.2 | - | 2, 3 | 567 | 5266336 | |
BC118604.1 | - | 2, 3 | 563 | 40109007 | ||
BC121816.1 | - | 2, 3 | 563 | 40109006 |