Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271856.1 | NP_001258785.1 | 2 | 103 | ||
NM_181711.3 | NP_859062.1 | 3 | 403 | |||
XM_005268691.3 | XP_005268748.1 | 3 | 1571 | |||
XM_011537996.1 | XP_011536298.1 | 2 | 1349 | |||
GenBank mRNA | BC035500.2 | - | 2 | 74 | 4422992 | |
BC052967.1 | - | 2 | 87 | |||
BC053666.1 | - | 3 | 384 | 6151168 | ||
HY035281.1 | - | 2 | 103 | |||
HY052304.1 | - | 3 | 404 |