Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPRIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034841.3 | NP_001030013.1 | 1 | 2124 | ||
NR_028028.1 | 2 | 1608 | ||||
GenBank mRNA | AK096327.1 | - | 2 | 1608 | ||
AK097274.1 | - | 1 | 1570 | |||
BC101749.1 | - | 1 | 1673 | 8069255 | ||
BC101751.1 | - | 1 | 1673 | 8069257 |